Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g/mol | C10H10O2 | 50 G
SDP
Catalog No. 5000301184
Encompass_Preferred
Phenyl methacrylate is a hydrophobic methacrylate monomer (CAS 2177-70-0) used as a comonomer or building block in polymer synthesis. The aromatic phenyl group imparts hydrophobicity, rigidity, and improved UV resistance, and the material has been reported for use in specialty optical and polymer applications such as light-defocusing plastic rods.
  • Hydrophobic methacrylate monomer suitable for copolymerization.
  • Aromatic phenyl group increases rigidity and UV stability.
  • Liquid, colorless to light yellow, convenient for handling and formulation.
  • Molecular weight 162.19 g/mol and formula C10H10O2 for stoichiometric calculations.
  • Store protected from light and under inert atmosphere to preserve quality.
Catalog No. 50-003-01184 Supplier Medchemexpress LLC Supplier No. HYW01721350G
Phenyl methacrylate is a hydrophobic methacrylate monomer (CAS 2177-70-0) used as a comonomer or building block in polymer synthesis. The aromatic phenyl group imparts hydrophobicity, rigidity, and improved UV resistance, and the material has been reported for use in specialty optical and polymer applications such as light-defocusing plastic rods.
  • Hydrophobic methacrylate monomer suitable for copolymerization.
  • Aromatic phenyl group increases rigidity and UV stability.
  • Liquid, colorless to light yellow, convenient for handling and formulation.
  • Molecular weight 162.19 g/mol and formula C10H10O2 for stoichiometric calculations.
  • Store protected from light and under inert atmosphere to preserve quality.