Learn More
Medchemexpress LLC Phenyl methacrylate | 2177-70-0 | MFCD00048117 | 162.19 g/mol | C10H10O2 | 500 G
SDP
Catalog No. 5000299522
Encompass_Preferred
Phenyl methacrylate is a hydrophobic methacrylate monomer (C10H10O2, MW 162.19 g/mol, CAS 2177-70-0) used in polymer synthesis and specialty materials such as light-defocusing plastic rods. It is supplied as a colorless to light-yellow liquid and is typically handled under inert atmosphere and protected from light.
  • Hydrophobic methacrylate monomer suitable for radical polymerization.
  • Used in polymer synthesis and specialty optical materials.
  • Colorless to light-yellow liquid at room temperature.
  • Density approximately 1.053 g/cm³.
  • Recommend storage at 4°C, protected from light, under nitrogen.
  • Available in multiple pack sizes, including 500 g.
Catalog No. 50-002-99522 Supplier Medchemexpress LLC Supplier No. HYW017213500G
Phenyl methacrylate is a hydrophobic methacrylate monomer (C10H10O2, MW 162.19 g/mol, CAS 2177-70-0) used in polymer synthesis and specialty materials such as light-defocusing plastic rods. It is supplied as a colorless to light-yellow liquid and is typically handled under inert atmosphere and protected from light.
  • Hydrophobic methacrylate monomer suitable for radical polymerization.
  • Used in polymer synthesis and specialty optical materials.
  • Colorless to light-yellow liquid at room temperature.
  • Density approximately 1.053 g/cm³.
  • Recommend storage at 4°C, protected from light, under nitrogen.
  • Available in multiple pack sizes, including 500 g.