Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl-p-benzoquinone, 95%

Catalog No. AC417381000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417381000 100 g Glass bottle
AC417380250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417381000 Supplier Thermo Scientific Chemicals Supplier No. 417381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 363-03-1 | C12H8O2 | 184.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-03-1
Molecular Formula C12H8O2
Molecular Weight (g/mol) 184.19
MDL Number MFCD00001599
InChI Key RLQZIECDMISZHS-UHFFFAOYSA-N
Synonym phenyl-p-benzoquinone, phenylquinone, phenylbenzoquinone, 2-phenyl-1,4-benzoquinone, 2-phenylbenzoquinone, p-benzoquinone, 2-phenyl, 2,5-cyclohexadiene-1,4-dione, 2-phenyl, 2-phenyl-p-benzoquinone, phenyl-1,4-benzoquinone, p-benzoquinone, phenyl
PubChem CID 9688
IUPAC Name 2-phenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=CC(=O)C(=C1)C1=CC=CC=C1

Specifications

Melting Point 111°C to 115°C
Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,benzene,petroleum ether;,very soluble in chloroform
Formula Weight 184.19
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material Phenyl-p-benzoquinone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-654-5
RTECSNumber DK6650000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.