Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl p-Tolyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. P174025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P174025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P174025G Supplier TCI America Supplier No. P174025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3699-01-2
Molecular Formula C13H12S
Molecular Weight (g/mol) 200.30
MDL Number MFCD00092378
InChI Key CPZFPNKPHBCUOB-UHFFFAOYSA-N
Synonym 4-(Phenylthio)toluene
PubChem CID 138011
IUPAC Name 1-methyl-4-phenylsulfanylbenzene
SMILES CC1=CC=C(C=C1)SC2=CC=CC=C2

Specifications

Boiling Point 312°C
Quantity 25 g
Formula Weight 200.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl p-Tolyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.