Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Phenyl(propyl)sulfane | ≥97% | CAS 874-79-3 | C9H12S | 5g

Catalog No. 5000984975
Encompass_Preferred

Phenyl(propyl)sulfane | ≥97% | CAS 874-79-3 | C9H12S | 5g

Catalog No. 50-009-84975 Supplier Aobchem Supplier No. A1645435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl(propyl)sulfane | ≥97% | CAS 874-79-3 | C9H12S | 5g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.