Learn More
Medchemexpress LLC Phenyl salicylate | 118-55-8 | 99.98% | 250 G
SDP
Catalog No. 5000414351
Encompass_Preferred
Phenyl salicylate (PS) exhibits antibacterial activity upon hydrolysis in the small intestine. It is commonly utilized as a nervous system inhibitor and intestinal preservative.
  • Exhibits antibacterial activity upon hydrolysis
  • Functions as a nervous system inhibitor
  • Acts as an intestinal preservative
Catalog No. 50-004-14351 Supplier Medchemexpress LLC Supplier No. 250GHYB1731
Phenyl salicylate (PS) exhibits antibacterial activity upon hydrolysis in the small intestine. It is commonly utilized as a nervous system inhibitor and intestinal preservative.
  • Exhibits antibacterial activity upon hydrolysis
  • Functions as a nervous system inhibitor
  • Acts as an intestinal preservative