Learn More
Medchemexpress LLC Phenyl salicylate | 118-55-8 | MFCD00002213 | 99.9% | 500 G
SDP
Catalog No. 5000412392
Encompass_Preferred
Phenyl salicylate (PS) exhibits antibacterial activity upon hydrolysis in the small intestine and is frequently employed as a nervous system inhibitor and intestinal preservative. It is utilized in the manufacturing of polymers, lacquers, adhesives, waxes, and polishes, and functions as an ultraviolet filter. This compound also serves as an analgesic, antipyretic, and antiseptic, with additional applications as an antioxidant and food stabilizer.
  • Exhibits antibacterial activity
  • Functions as a nervous system inhibitor
  • Acts as an intestinal preservative
  • Used in polymer, lacquer, adhesive, wax, and polish manufacturing
  • Serves as an ultraviolet filter
  • Offers pain relief as an analgesic
  • Reduces fever as an antipyretic
  • Possesses antiseptic properties
  • Functions as an antioxidant and food stabilizer
Catalog No. 50-004-12392 Supplier Medchemexpress LLC Supplier No. HYB1731500G
Phenyl salicylate (PS) exhibits antibacterial activity upon hydrolysis in the small intestine and is frequently employed as a nervous system inhibitor and intestinal preservative. It is utilized in the manufacturing of polymers, lacquers, adhesives, waxes, and polishes, and functions as an ultraviolet filter. This compound also serves as an analgesic, antipyretic, and antiseptic, with additional applications as an antioxidant and food stabilizer.
  • Exhibits antibacterial activity
  • Functions as a nervous system inhibitor
  • Acts as an intestinal preservative
  • Used in polymer, lacquer, adhesive, wax, and polish manufacturing
  • Serves as an ultraviolet filter
  • Offers pain relief as an analgesic
  • Reduces fever as an antipyretic
  • Possesses antiseptic properties
  • Functions as an antioxidant and food stabilizer