Learn More
U.S. Pharmacopeia Phenyl Salicylate Melting Point Standard, 118-55-8, MFCD00002213, 1g
Catalog No. NC0326700
Encompass
Molecular formula C13H10O3, Molecular weight 214.22, Melting point 104.72 - 111.74 °F (40.4 - 44.3 °C), Boiling point 579.2 - 582.8 °F (304 - 306 °C) at 760 mm Hg, Flash point 235.4 °F (113.0 °C) (CC), Synonyms: Salol, 2-phenoxycarbonylphenol
Catalog No. NC0326700 Supplier U.S. Pharmacopeia Supplier No. 1534209
Molecular formula C13H10O3, Molecular weight 214.22, Melting point 104.72 - 111.74 °F (40.4 - 44.3 °C), Boiling point 579.2 - 582.8 °F (304 - 306 °C) at 760 mm Hg, Flash point 235.4 °F (113.0 °C) (CC), Synonyms: Salol, 2-phenoxycarbonylphenol