Learn More
Bioworld PHENYL SEPAROPOREAGAROSE)
Supplier:  Bioworld 201810743
Encompass
PHENYL SEPAROPORE (AGAROSE) 4B-CL: Application: Hydrophobic chromatorgraphy. Specifications: Ligand: Phenyl group.; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% B...
Catalog No. 50-197-860