Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenyl SepharoseR High Per
Catalog No. 502904691
Encompass
Phenyl Sepharose(R) High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium designed for intermediate and polishing step purification steps when high reSOLUTIONon has priority.
Catalog No. 50-290-4691 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17108201
Phenyl Sepharose(R) High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium designed for intermediate and polishing step purification steps when high reSOLUTIONon has priority.