Learn More
Medchemexpress LLC Phenyl sulfate | 937-34-8 | 98% | 50 MG
SDP
Catalog No. 5000341836
Encompass_Preferred
Phenyl sulfate (Standard) is an orally active gut microbiota-derived metabolite, primarily used as an analytical standard for research and analytical applications. It has been identified as a disease marker in diabetic kidney disease, capable of inducing albuminuria and podocyte damage in experimental diabetes models. This analytical standard is suitable for use in qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Used as an analytical standard for research
  • Suitable for HPLC, GC, and MS applications
  • Functions as an orally active gut microbiota-derived metabolite
  • Identified as a disease marker in diabetic kidney disease
Catalog No. 50-003-41836 Supplier Medchemexpress LLC Supplier No. HY12844250MG
Phenyl sulfate (Standard) is an orally active gut microbiota-derived metabolite, primarily used as an analytical standard for research and analytical applications. It has been identified as a disease marker in diabetic kidney disease, capable of inducing albuminuria and podocyte damage in experimental diabetes models. This analytical standard is suitable for use in qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Used as an analytical standard for research
  • Suitable for HPLC, GC, and MS applications
  • Functions as an orally active gut microbiota-derived metabolite
  • Identified as a disease marker in diabetic kidney disease