Learn More
Medchemexpress LLC Phenyl sulfate | 937-34-8 | 99.5% | 1 ML
SDP
Catalog No. 5000341846
Encompass_Preferred
Phenyl sulfate is an orally active gut microbiota-derived metabolite. It induces albuminuria and podocyte damage in podocyte damage in experimental models of diabetes. It can be used as a disease marker in diabetic kidney disease. This product is for research use only and not sold to patients.
  • Orally active gut microbiota-derived metabolite
  • Induces albuminuria and podocyte damage in experimental models of diabetes
  • Functions as a disease marker in diabetic kidney disease
  • For research use only
Catalog No. 50-003-41846 Supplier Medchemexpress LLC Supplier No. HY12844210MM/1ML
Phenyl sulfate is an orally active gut microbiota-derived metabolite. It induces albuminuria and podocyte damage in podocyte damage in experimental models of diabetes. It can be used as a disease marker in diabetic kidney disease. This product is for research use only and not sold to patients.
  • Orally active gut microbiota-derived metabolite
  • Induces albuminuria and podocyte damage in experimental models of diabetes
  • Functions as a disease marker in diabetic kidney disease
  • For research use only