Learn More
Medchemexpress LLC Phenyl sulfate | 937-34-8 | 99.5% | 25 MG
SDP
Catalog No. 5000343067
Encompass_Preferred
Phenyl sulfate is an orally active gut microbiota-derived metabolite. It induces albuminuria and podocyte damage in experimental models of diabetes and can be used as a disease marker in diabetic kidney disease.
  • Orally active gut microbiota-derived metabolite
  • Induces albuminuria and podocyte damage in experimental models
  • Used as a disease marker in diabetic kidney disease
Catalog No. 50-003-43067 Supplier Medchemexpress LLC Supplier No. 25MGHY128442
Phenyl sulfate is an orally active gut microbiota-derived metabolite. It induces albuminuria and podocyte damage in experimental models of diabetes and can be used as a disease marker in diabetic kidney disease.
  • Orally active gut microbiota-derived metabolite
  • Induces albuminuria and podocyte damage in experimental models
  • Used as a disease marker in diabetic kidney disease