Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Vinyl Sulfide (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. P128725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P128725G 25 g
P12875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P128725G Supplier TCI America Supplier No. P128725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1822-73-7
Molecular Formula C8H8S
Molecular Weight (g/mol) 136.21
MDL Number MFCD00009646
InChI Key GMPDOIGGGXSAPL-UHFFFAOYSA-N
Synonym phenyl vinyl sulfide, phenylthioethene, phenyl vinyl sulphide, phenylthio ethylene, vinylthiobenzene, benzene, ethenylthio, vinyl phenyl sulfide, sulfide, phenyl vinyl, vinylsulfanyl benzene, phenyl vinyl thioether
PubChem CID 74572
IUPAC Name (ethenylsulfanyl)benzene
SMILES C=CSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
UN Number 1993
Formula Weight 136.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Vinyl Sulfide (stabilized with TBC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.