Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetaldehyde 85%, Thermo Scientific™

Catalog No. p-181937
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148181000 100 g
AC148185000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148181000 Supplier Thermo Scientific Chemicals Supplier No. 148181000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Acidity 5mg KOH/g max.
Color Colorless to Yellow
Density 1.54
Infrared Spectrum Authentic
Refractive Index 1.5300 to 1.5420 (20°C, 589nm)
Linear Formula C6H5CH2CHO
Quantity 100 g
Beilstein 07, 292
Specific Gravity 1.044 to 1.064 (20°C)
Merck Index 15, 7377
Formula Weight 120.15
Percent Purity ≥80%
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde, 85%
Show More Show Less
EINECSNumber 204-574-5
RTECSNumber CY1420000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.