Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenylacetaldehyde 95 fCC1KG
Catalog No. 502897907
Encompass
Phenylacetaldehyde is an important aroma volatile found in tomato and roses. It has also been identified in potato roasted cocoa beans and honey. Phenylacetaldehyde is also a potent moth attractant.
Catalog No. 50-289-7907 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2874071KGK
Phenylacetaldehyde is an important aroma volatile found in tomato and roses. It has also been identified in potato roasted cocoa beans and honey. Phenylacetaldehyde is also a potent moth attractant.