Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetaldehyde, 99.5%, For GC analysis, MP Biomedicals™

Catalog No. ICN15614580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15614580 100 mL
MP215614590 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15614580 Supplier MP Biomedicals, Inc Supplier No. 0215614580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylacetaldehyde ≥95%, Clear Liquid

  • It is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions.
  • It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages.
  • It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin.
  • Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.
  • A naturally occurring aromatic compound. Contains varying amounts of poly(styrene oxide).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.027g/mL
Assay Percent Range 0.995
Quantity 100 mL
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
Formula Weight 120.15
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde, For GC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.