Learn More
Sigma Organic Chemistry phenylacetyl aminooxy a 2 | 25mg | MFCD04037285
Supplier: Sigma Organic Chemistry CDS01285825MG
            phenylacetyl aminooxy a  2, 25mg
About this Item:
MDL #: MFCD04037285
Molecular Weight: 209.2
UNSPSC Code: 12352200
Chemical Formula: C10H11NO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.