Learn More
Cayman Chemical Phenylacetyl L-Glutamine, Purity: ≥95%, CAS: 28047-15-6, 100 mg
SDP
Catalog No. 502968248
Encompass_Preferred
An end product of phenylalanine metabolism in humans and primates
Catalog No. 50-296-8248 Supplier Cayman Chemical Supplier No. 16724100 MG
An end product of phenylalanine metabolism in humans and primates