Learn More
Cayman Chemical Phenylacetyl L-Glutamine, Purity: ≥95%, CAS: 28047-15-6, 25 mg
SDP
Catalog No. 502965323
Encompass_Preferred
An end product of phenylalanine metabolism in humans and primates
Catalog No. 50-296-5323 Supplier Cayman Chemical Supplier No. 1672425 MG
An end product of phenylalanine metabolism in humans and primates