Learn More
Cayman Chemical Phenylacetylglycine, Purity: ≥98%, CAS: 500-98-1, 50 mg
SDP
Catalog No. 502911797
Encompass_Preferred
A glycine conjugate of phenylacetate; decreases hypoxia-induced apoptosis in neonatal mouse cardiomyocytes at 100 µM; urine and plasma levels are increased in an amiodarone-induced rat model of phospholipidosis; reduces myocardial infarct size in a mouse model of ischemia-reperfusion injury
Catalog No. 50-291-1797 Supplier Cayman Chemical Supplier No. 3636850 MG
A glycine conjugate of phenylacetate; decreases hypoxia-induced apoptosis in neonatal mouse cardiomyocytes at 100 µM; urine and plasma levels are increased in an amiodarone-induced rat model of phospholipidosis; reduces myocardial infarct size in a mouse model of ischemia-reperfusion injury