Learn More
Medchemexpress LLC Phenylalanine-4'-azobenzene HCl | 2137036-84-9 | 99.3% | 305.76 g/mol | C15H16ClN3O2 | 100 MG
SDP
Catalog No. 5000283047
Encompass_Preferred
Phenylalanine-4′-azobenzene HCl is an amino-acid-derived azobenzene hydrochloride supplied as a light yellow to brown solid for research use. The compound combines a phenylalanine backbone with a photoresponsive azobenzene group, making it useful as a probe or synthetic intermediate in photochemistry and biochemical studies. It is soluble in DMSO and requires sealed, dry, low-temperature storage.
Catalog No. 50-002-83047 Supplier Medchemexpress LLC Supplier No. HYW141859100MG
Phenylalanine-4′-azobenzene HCl is an amino-acid-derived azobenzene hydrochloride supplied as a light yellow to brown solid for research use. The compound combines a phenylalanine backbone with a photoresponsive azobenzene group, making it useful as a probe or synthetic intermediate in photochemistry and biochemical studies. It is soluble in DMSO and requires sealed, dry, low-temperature storage.