Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylalanine-4'-azobenzene HCl | 2137036-84-9 | MFCD30187602 | 99.3% | 305.76 | C15H16ClN3O2 | 50 MG
SDP

Catalog No. 5000283792
Encompass_Preferred

Phenylalanine-4'-azobenzene HCl is an amino-acid-derived azo compound supplied as the hydrochloride salt for research use. It is provided as a solid with documented molecular formula and molecular weight, and is intended for laboratory studies involving photoresponsive amino-acid analogs.

  • Amino-acid-derived azo compound suitable for research use.
  • CAS number 2137036-84-9 for unambiguous identification.
  • Molecular weight 305.76 g/mol.
  • Purity 99.3%.
  • Available in small research quantities such as 50 MG.
  • Store sealed at -20°C; follow low-temperature storage guidelines for solutions.

Catalog No. 50-002-83792 Supplier Medchemexpress LLC Supplier No. HYW14185950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylalanine-4'-azobenzene HCl is an amino-acid-derived azo compound supplied as the hydrochloride salt for research use. It is provided as a solid with documented molecular formula and molecular weight, and is intended for laboratory studies involving photoresponsive amino-acid analogs.

  • Amino-acid-derived azo compound suitable for research use.
  • CAS number 2137036-84-9 for unambiguous identification.
  • Molecular weight 305.76 g/mol.
  • Purity 99.3%.
  • Available in small research quantities such as 50 MG.
  • Store sealed at -20°C; follow low-temperature storage guidelines for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.