Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylalanine, N-[(1,1-dimethylethoxy)carbonyl]-N,β,β-trimethyl- | 676487-35-7 | 99.1% | 307.38 | 250 MG
SDP

Catalog No. 5000402315
Encompass_Preferred

(Rac)-Taltobulin intermediate-1 is an intermediate used in the synthesis of Taltobulin. Taltobulin is a common toxin component in ADC preparation and acts as a powerful tubulin inhibitor. It works by disrupting tubulin polymerization, inducing mitotic arrest, and promoting apoptosis.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-02315 Supplier Medchemexpress LLC Supplier No. 250MGHY41045
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Taltobulin intermediate-1 is an intermediate used in the synthesis of Taltobulin. Taltobulin is a common toxin component in ADC preparation and acts as a powerful tubulin inhibitor. It works by disrupting tubulin polymerization, inducing mitotic arrest, and promoting apoptosis.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.