Learn More
Medchemexpress LLC Phenylalanine, N-[(1,1-dimethylethoxy)carbonyl]-N, β,β-trimethyl- | 676487-35-7 | 99.1% | 307.38 | 50 MG
SDP
Catalog No. 5000404071
Encompass_Preferred
(Rac)-Taltobulin intermediate-1 is an intermediate used in the synthesis of Taltobulin. Taltobulin is identified as a common toxin component in Antibody-Drug Conjugate (ADC) preparation and acts as a powerful inhibitor of tubulin. Its mechanism of action involves disrupting tubulin polymerization, leading to mitotic arrest and inducing apoptosis.
  • Intermediate in the synthesis of Taltobulin.
  • A component of ADC preparation.
  • A potent tubulin inhibitor.
  • Disrupts tubulin polymerization, induces mitotic arrest, and causes apoptosis.
  • Available as solid or in solution.
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
Catalog No. 50-004-04071 Supplier Medchemexpress LLC Supplier No. HY4104550MG
(Rac)-Taltobulin intermediate-1 is an intermediate used in the synthesis of Taltobulin. Taltobulin is identified as a common toxin component in Antibody-Drug Conjugate (ADC) preparation and acts as a powerful inhibitor of tubulin. Its mechanism of action involves disrupting tubulin polymerization, leading to mitotic arrest and inducing apoptosis.
  • Intermediate in the synthesis of Taltobulin.
  • A component of ADC preparation.
  • A potent tubulin inhibitor.
  • Disrupts tubulin polymerization, induces mitotic arrest, and causes apoptosis.
  • Available as solid or in solution.
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).