Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylammonium iodide

Catalog No. 80591225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-591-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-591-225G Supplier Sigma Aldrich Supplier No. 80591225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.Product of Greatcell Solar(R)

Specifications

CAS 45497-73-2
Molecular Formula C6H8IN
Linear Formula C6H8IN
Quantity 25 g
MDL Number MFCD21607646
Synonym Aniline hydriodide; Anilinium iodide; Benzenamine hydriodide; Benzenaminium iodide; Greatcell Solar™
Molecular Weight (g/mol) 221.04
Percent Purity ≥98% (H-NMR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.