Learn More
Cayman Chemical Phenylarsine Oxide, Purity: ≥95%, CAS: 637-03-6, 1 g
SDP
Catalog No. 502909734
Encompass_Preferred
An organoarsenic compound; inhibits insulin-induced PP1 activity and Glut4 translocation in rat adipocytes at 5 µM; completely inhibits insulin-induced PI3K activation and glucose transport in 3T3-L1 adipocytes at 30 and 50 µM, respectively; inhibits potassium chloride-induced exocytosis and induces mitochondrial depolarization in isolated rat brain synaptosomes; reduces LPS-induced neutrophil infiltration in BALF and carrageenan-induced paw edema in rats at 0.3 and 1 mg/kg, i.p
Catalog No. 50-290-9734 Supplier Cayman Chemical Supplier No. 355251 G
An organoarsenic compound; inhibits insulin-induced PP1 activity and Glut4 translocation in rat adipocytes at 5 µM; completely inhibits insulin-induced PI3K activation and glucose transport in 3T3-L1 adipocytes at 30 and 50 µM, respectively; inhibits potassium chloride-induced exocytosis and induces mitochondrial depolarization in isolated rat brain synaptosomes; reduces LPS-induced neutrophil infiltration in BALF and carrageenan-induced paw edema in rats at 0.3 and 1 mg/kg, i.p