Learn More
Medchemexpress LLC Phenylbiguanide | 102-02-3 | MFCD00179077 | 98.0% | 177.21 g/mol | C8H11N5 | 1 ML
SDP
Catalog No. 5000212810
Encompass_Preferred
Phenylbiguanide is a selective 5-HT3 receptor agonist used in pharmacology and neuroscience research to probe 5-HT3-mediated responses. It is supplied as a pre-made 10 mM solution in DMSO (1 mL) and is also available in solid forms, offering defined purity and stability for reproducible in vitro and in vivo preparation.
  • Selective 5-HT3 receptor agonist with EC50 ≈ 3.0 μM.
  • Offered as a 10 mM solution in DMSO and in solid pack sizes.
  • High purity (98.0%) for reliable experimental results.
  • Soluble in DMSO and water for flexible formulation options.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-12810 Supplier Medchemexpress LLC Supplier No. HY10133110MM 1ML
Phenylbiguanide is a selective 5-HT3 receptor agonist used in pharmacology and neuroscience research to probe 5-HT3-mediated responses. It is supplied as a pre-made 10 mM solution in DMSO (1 mL) and is also available in solid forms, offering defined purity and stability for reproducible in vitro and in vivo preparation.
  • Selective 5-HT3 receptor agonist with EC50 ≈ 3.0 μM.
  • Offered as a 10 mM solution in DMSO and in solid pack sizes.
  • High purity (98.0%) for reliable experimental results.
  • Soluble in DMSO and water for flexible formulation options.
  • Stable under recommended storage conditions for long-term use.