Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylbiguanide | 55-57-2 | MFCD00035040 | 100.0% | 213.67 g/mol | C8H12ClN5 | 100 G
SDP

Catalog No. 5000212811
Encompass_Preferred

Phenylbiguanide hydrochloride is a research-grade small molecule used as a selective agonist of the 5-HT3 receptor in pharmacology and neuronal signaling studies. It exhibits an EC50 of approximately 3.0 μM and has documented solubility and stability data suitable for laboratory use.

  • Selective 5-HT3 receptor agonist (EC50 ≈ 3.0 μM).
  • Chemical identity: CAS 55-57-2; formula C8H12ClN5; molecular weight 213.67 g/mol.
  • High purity for research applications (COA reports 99.96%).
  • Soluble in DMSO (≈40 mg/mL); formulations reported for in vivo dosing.
  • Stable as solid at room temperature; defined solvent storage conditions available.
  • Suitable for GPCR and neuronal signaling assays and related pharmacology studies.

Catalog No. 50-002-12811 Supplier Medchemexpress LLC Supplier No. HY101331A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylbiguanide hydrochloride is a research-grade small molecule used as a selective agonist of the 5-HT3 receptor in pharmacology and neuronal signaling studies. It exhibits an EC50 of approximately 3.0 μM and has documented solubility and stability data suitable for laboratory use.

  • Selective 5-HT3 receptor agonist (EC50 ≈ 3.0 μM).
  • Chemical identity: CAS 55-57-2; formula C8H12ClN5; molecular weight 213.67 g/mol.
  • High purity for research applications (COA reports 99.96%).
  • Soluble in DMSO (≈40 mg/mL); formulations reported for in vivo dosing.
  • Stable as solid at room temperature; defined solvent storage conditions available.
  • Suitable for GPCR and neuronal signaling assays and related pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.