Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylbiguanide hydrochloride | 55-57-2 | 100.0% | 213.67 g/mol | C8H12ClN5 | 1 ML
SDP

Catalog No. 5000211218
Encompass_Preferred

Phenylbiguanide hydrochloride is a research-grade 5-HT3 receptor selective agonist used in receptor pharmacology and electrophysiology. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms for preparation. CAS number 55-57-2; reported EC50 ≈ 3.0 ± 0.1 μM.

  • Selective 5-HT3 receptor agonist with an EC50 of 3.0 ± 0.1 μM.
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High purity suitable for research applications.
  • Molecular weight 213.67 g/mol; molecular formula C8H12ClN5.
  • Formulation and solubility guidance provided for in vitro and in vivo studies.

Catalog No. 50-002-11218 Supplier Medchemexpress LLC Supplier No. HY101331A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylbiguanide hydrochloride is a research-grade 5-HT3 receptor selective agonist used in receptor pharmacology and electrophysiology. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms for preparation. CAS number 55-57-2; reported EC50 ≈ 3.0 ± 0.1 μM.

  • Selective 5-HT3 receptor agonist with an EC50 of 3.0 ± 0.1 μM.
  • Ready-to-use 10 mM solution in DMSO for in vitro assays.
  • High purity suitable for research applications.
  • Molecular weight 213.67 g/mol; molecular formula C8H12ClN5.
  • Formulation and solubility guidance provided for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.