Learn More
Medchemexpress LLC Phenylbiguanide hydrochloride | 55-57-2 | MFCD00035040 | 98.0% | 213.67 g/mol | C8H12ClN5 | 50 G
SDP
Catalog No. 5000211505
Encompass_Preferred
Phenylbiguanide hydrochloride is a research-grade small molecule that functions as a selective agonist of the 5-HT3 receptor. Supplied as the hydrochloride salt, it is used in in vitro pharmacology and neuroscience studies to characterize 5-HT3-mediated signaling and downstream GPCR responses.
  • Selective 5-HT3 receptor agonist with reported EC50 = 3.0 ± 0.1 μM.
  • Suitable for pharmacology and neuroscience research applications.
  • Molecular weight 213.67 g/mol; CAS number 55-57-2.
  • Available as a bulk reagent; common pack sizes include 50 g.
  • Research grade, for in vitro use only; not for human or veterinary use.
Catalog No. 50-002-11505 Supplier Medchemexpress LLC Supplier No. HY101331A50G
Phenylbiguanide hydrochloride is a research-grade small molecule that functions as a selective agonist of the 5-HT3 receptor. Supplied as the hydrochloride salt, it is used in in vitro pharmacology and neuroscience studies to characterize 5-HT3-mediated signaling and downstream GPCR responses.
  • Selective 5-HT3 receptor agonist with reported EC50 = 3.0 ± 0.1 μM.
  • Suitable for pharmacology and neuroscience research applications.
  • Molecular weight 213.67 g/mol; CAS number 55-57-2.
  • Available as a bulk reagent; common pack sizes include 50 g.
  • Research grade, for in vitro use only; not for human or veterinary use.