Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B085725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B085725G 25 g
B08575G 5 g
B0857-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B085725G Supplier TCI America Supplier No. B085725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid

Specifications

Melting Point 216°C
Quantity 25 g
SMILES OB(O)C1=CC=CC=C1
Formula Weight 121.93
Physical Form Crystalline Powder
Chemical Name or Material Phenylboronic Acid (contains varying amounts of Anhydride)
EINECSNumber (3)-4668
RTECSNumber CY8575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.