Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Phenylboronic acid, 98-80-6, MFCD00002103, 100g

Catalog No. 501472375
Encompass_Preferred

Molecular Formula C6H7BO2, Purity 98%, Molecular Weight 121.93, Melting Point 219°

Catalog No. 50-147-2375 Supplier Matrix Scientific Supplier No. 004911100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C6H7BO2, Purity 98%, Molecular Weight 121.93, Melting Point 219°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.