Learn More
Med Vet International Phenylbutazone Powder [Bute] Citrus, 2.2 Lbs
Catalog No. 502828876
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
A synthetic, non-hormonal, anti-inflammatory agent that is indicated for the relief of inflammatory conditions associated with the musculoskeletal system in horses. This highly palatable citrus-flavored powder is easily top-dressed on the horses ration, and is dosed at 0.6-1.2 oz. per 500 lbs. of body weight. Each 10 grams of powder contains 1 gram of phenylbutazone
Catalog No. 50-282-8876 Supplier Med Vet International Supplier No. RXBUTEPOWDER2
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
A synthetic, non-hormonal, anti-inflammatory agent that is indicated for the relief of inflammatory conditions associated with the musculoskeletal system in horses. This highly palatable citrus-flavored powder is easily top-dressed on the horses ration, and is dosed at 0.6-1.2 oz. per 500 lbs. of body weight. Each 10 grams of powder contains 1 gram of phenylbutazone