Learn More
Medchemexpress LLC HY-B0230 500mg Medchemexpress, Phenylbutazone CAS:50-33-9 Purity:>98%
SDP
Catalog No. 502082417
Encompass_Preferred
Medchemexpress, HY-B0230 500mg Phenylbutazone CAS:50-33-9 Phenylbutazone is an efficient reducing cofactor for the peroxidase activity of prostaglandin H synthase (PHS). Phenylbutazone, a hepatotoxin, is a nonsteroidal anti-inflammatory drug (NSAID). Phenylbutazone induces muscle blind-like protein 1 (MBNL1) expression and has the potential for ankylosing spondylitis research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2417 Supplier Medchemexpress LLC Supplier No. HYB0230500MG
Medchemexpress, HY-B0230 500mg Phenylbutazone CAS:50-33-9 Phenylbutazone is an efficient reducing cofactor for the peroxidase activity of prostaglandin H synthase (PHS). Phenylbutazone, a hepatotoxin, is a nonsteroidal anti-inflammatory drug (NSAID). Phenylbutazone induces muscle blind-like protein 1 (MBNL1) expression and has the potential for ankylosing spondylitis research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.