Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylbutazone (diphenyl-d10) | 1219794-69-0 | 98.8% | 318.44 g/mol | C19H10D10N2O2 | 10 MG
SDP

Catalog No. 5000297411
Encompass_Preferred

Phenylbutazone (diphenyl-d10) is a deuterium-labeled analogue of phenylbutazone provided for research and analytical applications. It is intended as a stable isotope internal standard for mass spectrometry and quantitative analysis, aiding accurate compound identification and quantification.

  • Deuterium-labeled internal standard for analytical workflows.
  • High purity suitable for quantitative mass spectrometry.
  • CAS number 1219794-69-0 for unambiguous identification.
  • Molecular weight 318.44 g/mol.
  • Provided as a 10 mg quantity for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-97411 Supplier Medchemexpress LLC Supplier No. HYB0230S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylbutazone (diphenyl-d10) is a deuterium-labeled analogue of phenylbutazone provided for research and analytical applications. It is intended as a stable isotope internal standard for mass spectrometry and quantitative analysis, aiding accurate compound identification and quantification.

  • Deuterium-labeled internal standard for analytical workflows.
  • High purity suitable for quantitative mass spectrometry.
  • CAS number 1219794-69-0 for unambiguous identification.
  • Molecular weight 318.44 g/mol.
  • Provided as a 10 mg quantity for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.