Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylbutazone, MP Biomedicals™

Catalog No. ICN15356780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15356780 100 g
ICN15356725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15356780 Supplier MP Biomedicals, Inc Supplier No. 0215356780

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-33-9
Molecular Formula C19H20N2O2
Molecular Weight (g/mol) 308.38
MDL Number MFCD00005500
InChI Key VYMDGNCVAMGZFE-UHFFFAOYSA-N
Synonym phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion
PubChem CID 4781
ChEBI CHEBI:48574
IUPAC Name 4-butyl-1,2-diphenylpyrazolidine-3,5-dione
SMILES CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 100 g
Formula Weight 308.37
Physical Form Solid
Chemical Name or Material Phenylbutazone
EINECSNumber 200-029-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.