Learn More
Medchemexpress LLC Phenylbutyrate-d11 sodium | 1392208-11-5 | MFCD30742904 | 98.0% | 197.2 g/mol | C10D11NaO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15654S10MG
Encompass_Preferred
Phenylbutyrate-d11 sodium is the deuterium-labeled sodium salt of 4-phenylbutyrate supplied as a small research reagent for use as a stable-isotope standard, tracer, or analytical reference in mass spectrometry and pharmacokinetic studies.
  • High isotopic labeling (d11) suitable for tracer and quantitation studies.
  • Provided as the sodium salt for improved aqueous handling.
  • Small 10 mg pack convenient for analytical standards and method development.
  • Suitable as an internal standard in LC-MS/MS and other analytical workflows.
  • Accompanied by safety and product documentation for traceability and safe handling.
Catalog No. 50-002-87781