Learn More
Strem, An Ascensus Company CAS# 644-97-3. 50g. Phenyldichlorophosphine, 97%. MFCD00000528
SDP
Catalog No. 5090110285
Encompass
CAS# 644-97-3. 50g. Phenyldichlorophosphine, 97%. MFCD00000528. Molecular Weight: 178.99. Molecular Formula: C6H5PCl2. Color/form: yellow liq. Strem# 15-4150. http://www.strem.com/catalog/v/15-4150/
Catalog No. 50-901-10285 Supplier Strem, An Ascensus Company Supplier No. 15415050G
CAS# 644-97-3. 50g. Phenyldichlorophosphine, 97%. MFCD00000528. Molecular Weight: 178.99. Molecular Formula: C6H5PCl2. Color/form: yellow liq. Strem# 15-4150. http://www.strem.com/catalog/v/15-4150/