Learn More
Medchemexpress LLC Phenylephrine hydrochloride | 61-76-7 | MFCD00012605 | 99.99% | 100 MG
SDP
Catalog No. 502031080
Encompass_Preferred
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87, and 4.70 for α1D, α1B, and α1A receptors, respectively. It promotes cardiac fibroblast proliferation by activating CaN and evoking NFAT3 nuclear translocation. This compound also activates p38-MAPK isoforms and SAPKs/JNKs, leading to increased heart contractility and enhanced alveolar fluid clearance. It is for research use only.
  • Selective α1-adrenoceptor agonist
  • Promotes cardiac fibroblast proliferation
  • Activates CaN and evokes NFAT3 nuclear translocation
  • Increases contractility of the heart
  • Activates SAPKs/JNKs in neonatal ventricular myocytes
  • Increases alveolar fluid clearance
  • Accelerates absorption of pulmonary edema
Catalog No. 50-203-1080 Supplier Medchemexpress LLC Supplier No. HYB0471100MG
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist with pKis of 5.86, 4.87, and 4.70 for α1D, α1B, and α1A receptors, respectively. It promotes cardiac fibroblast proliferation by activating CaN and evoking NFAT3 nuclear translocation. This compound also activates p38-MAPK isoforms and SAPKs/JNKs, leading to increased heart contractility and enhanced alveolar fluid clearance. It is for research use only.
  • Selective α1-adrenoceptor agonist
  • Promotes cardiac fibroblast proliferation
  • Activates CaN and evokes NFAT3 nuclear translocation
  • Increases contractility of the heart
  • Activates SAPKs/JNKs in neonatal ventricular myocytes
  • Increases alveolar fluid clearance
  • Accelerates absorption of pulmonary edema