Learn More
Medchemexpress LLC Phenylephrine (hydrochloride) | 61-76-7 | 99.9% | 10 MM 1 ML
SDP
Catalog No. 5000405285
Encompass_Preferred
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist. It acts as an agonist for α1D, α1B, and α1A receptors, promoting cardiac fibroblast proliferation and activating the Ca(2+)/CaN/NFAT pathway, which mediates phenylephrine-induced cardiac fibroblast proliferation.
  • Selective α1-adrenoceptor agonist
  • Targets adrenergic receptor and endogenous metabolite
  • Suitable for cardiovascular disease, endocrinology, and cancer research
  • Promotes cardiac fibroblast proliferation
  • Activates CaN and evokes NFAT3 nuclear translocation
  • Used in laboratory chemicals and substance manufacturing
Catalog No. 50-004-05285 Supplier Medchemexpress LLC Supplier No. HYB047110MM/1ML
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist. It acts as an agonist for α1D, α1B, and α1A receptors, promoting cardiac fibroblast proliferation and activating the Ca(2+)/CaN/NFAT pathway, which mediates phenylephrine-induced cardiac fibroblast proliferation.
  • Selective α1-adrenoceptor agonist
  • Targets adrenergic receptor and endogenous metabolite
  • Suitable for cardiovascular disease, endocrinology, and cancer research
  • Promotes cardiac fibroblast proliferation
  • Activates CaN and evokes NFAT3 nuclear translocation
  • Used in laboratory chemicals and substance manufacturing