Learn More
Medchemexpress LLC Phenylephrine hydrochloride | 61-76-7 | 99.99% | 50 MG
SDP
Catalog No. 5000405405
Encompass_Preferred
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist that promotes cardiac fibroblast proliferation via the Ca(2+)/CaN/NFAT pathway. It rapidly activates p38-MAPK isoforms, increases heart contractility, and activates SAPKs/JNKs in neonatal ventricular myocytes. It also increases alveolar fluid clearance and accelerates the absorption of pulmonary edema.
  • Selective α1-adrenoceptor agonist
  • Promotes cardiac fibroblast proliferation
  • Activates CaN, evoking NFAT3 nuclear translocation
  • Mediates cardiac fibroblast proliferation via Ca(2+)/CaN/NFAT pathway
  • Causes rapid activation of p38-MAPK isoforms
  • Increases heart contractility
  • Activates SAPKs/JNKs in neonatal ventricular myocytes
  • Can increase alveolar fluid clearance
  • Accelerates absorption of pulmonary edema
Catalog No. 50-004-05405 Supplier Medchemexpress LLC Supplier No. HYB047150MG
Phenylephrine hydrochloride is a selective α1-adrenoceptor agonist that promotes cardiac fibroblast proliferation via the Ca(2+)/CaN/NFAT pathway. It rapidly activates p38-MAPK isoforms, increases heart contractility, and activates SAPKs/JNKs in neonatal ventricular myocytes. It also increases alveolar fluid clearance and accelerates the absorption of pulmonary edema.
  • Selective α1-adrenoceptor agonist
  • Promotes cardiac fibroblast proliferation
  • Activates CaN, evoking NFAT3 nuclear translocation
  • Mediates cardiac fibroblast proliferation via Ca(2+)/CaN/NFAT pathway
  • Causes rapid activation of p38-MAPK isoforms
  • Increases heart contractility
  • Activates SAPKs/JNKs in neonatal ventricular myocytes
  • Can increase alveolar fluid clearance
  • Accelerates absorption of pulmonary edema