Learn More
U.S. Pharmacopeia Phenylephrine Hydrochloride, 61-76-7, MFCD00012605, 200mg
Catalog No. NC9637739
Encompass
Molecular formula C9H13NO2 . HCl, Molecular weight 203.67, Melting point 284 - 293 °F (140 - 145 °C), Synonyms: m-Synephrine hydrochloride
Catalog No. NC9637739 Supplier U.S. Pharmacopeia Supplier No. 1533002
Molecular formula C9H13NO2 . HCl, Molecular weight 203.67, Melting point 284 - 293 °F (140 - 145 °C), Synonyms: m-Synephrine hydrochloride