Learn More
U.S. Pharmacopeia Phenylephrine Related Compound F, 1007885-60-0, MFCD29472421, 30mg
Catalog No. NC1667626
Encompass
Molecular formula C10H13NO2.HCl.H20, Molecular weight 233.7, Melting point/freezing point 458.6 - 462.2 °F (237 - 239 °C), Chemical name: (R)-2-methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol hydrochloride hydrate
Catalog No. NC1667626 Supplier U.S. Pharmacopeia Supplier No. 1533261
Molecular formula C10H13NO2.HCl.H20, Molecular weight 233.7, Melting point/freezing point 458.6 - 462.2 °F (237 - 239 °C), Chemical name: (R)-2-methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol hydrochloride hydrate