Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylethyl Alcohol, 98%, Spectrum™ Chemical
SDP

Catalog No. 18612303 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 kg
15.9 kg
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-303 15.9 kg Amber Glass Bottle
18-612-301 100 g Amber Glass Bottle
18-612-302 200 L Amber Glass Bottle
18-612-300 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-303 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P105715.9KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identifiants chimiques

CAS 60-12-8

Spécifications

CAS Max % 1
Boiling Point 218°C (424.4°F)
Packaging Amber Glass Bottle
Quantity 15.9 kg
Specific Gravity 1.017 - 1.019
Molecular Weight (g/mol) 122.17
Percent Purity 0.98
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Phenylethyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.