Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-phenylethyl β-D-glucoside | 18997-54-1 | MFCD00270030 | 97.0% | 284.31 g/mol | C14H20O6 | 25 MG
SDP

Catalog No. 5000285162
Encompass_Preferred

β-Phenylethyl β-D-glucoside is a naturally occurring glycoside supplied for laboratory research applications. The substance is provided as a powder and intended for research use only; analytical data from the manufacturer indicate high purity suitable for experimental studies.

  • For research use only.
  • Purity: 97.0%.
  • Chemical formula C14H20O6.
  • Molecular weight 284.31 g/mol.
  • Appearance: powder.
  • Available in small-scale packaging such as 25 MG.
  • Synonym: phenylethyl β-D-glucopyranoside.

Catalog No. 50-002-85162 Supplier Medchemexpress LLC Supplier No. HYW35714025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Phenylethyl β-D-glucoside is a naturally occurring glycoside supplied for laboratory research applications. The substance is provided as a powder and intended for research use only; analytical data from the manufacturer indicate high purity suitable for experimental studies.

  • For research use only.
  • Purity: 97.0%.
  • Chemical formula C14H20O6.
  • Molecular weight 284.31 g/mol.
  • Appearance: powder.
  • Available in small-scale packaging such as 25 MG.
  • Synonym: phenylethyl β-D-glucopyranoside.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.