Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

βPhenylethylamine HCl, MP Biomedicals

Catalog No. ICN217380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217380 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217380 Supplier MP Biomedicals, Inc Supplier No. 0521738025

Chemical Identifiers

CAS 156-28-5
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
InChI Key SKHIBNDAFWIOPB-UHFFFAOYSA-N
Synonym 2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d
PubChem CID 9075
IUPAC Name 2-phenylethanamine;hydrochloride
SMILES C1=CC=C(C=C1)CCN.Cl

Specifications

Quantity 25 g
Formula Weight 157.64
Chemical Name or Material Beta-Phenylethylamine HCL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.