Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylfluorone 97.0+%, TCI America™
SDP

Catalog No. T04505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04505G 5 g
T04501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04505G Supplier TCI America Supplier No. T04505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 975-17-7
Molecular Formula C19H12O5
Molecular Weight (g/mol) 320.3
MDL Number MFCD00005048
InChI Key YDCFOUBAMGLLKA-UHFFFAOYSA-N
Synonym phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone
PubChem CID 70420
IUPAC Name 2,6,7-trihydroxy-9-phenylxanthen-3-one
SMILES C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O

Specifications

Quantity 5 g
Formula Weight 320.30
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylfluorone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.