Learn More
Medchemexpress LLC Phenylglyoxylic acid | 611-73-4 | 99.5% | 150.13 | 50 MG
SDP
Catalog No. 5000453988
Encompass_Preferred
Phenylglyoxylic acid (Standard) is the analytical standard of Phenylglyoxylic acid. It is intended for research and analytical applications. It is a metabolite of ethylbenzene and styrene (EB/S) and can be used as a biomarker of exposure to EB/S in humans. It can also be used as a drug intermediate for the synthesis of antineoplastic compounds.
  • Analytical standard for research and analytical applications
  • Metabolite of ethylbenzene and styrene (EB/S)
  • Can be used as a biomarker of exposure to EB/S
  • Drug intermediate for the synthesis of antineoplastic compounds
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-53988 Supplier Medchemexpress LLC Supplier No. HYW010255R50MG
Add to Cart
Add to Cart
Phenylglyoxylic acid (Standard) is the analytical standard of Phenylglyoxylic acid. It is intended for research and analytical applications. It is a metabolite of ethylbenzene and styrene (EB/S) and can be used as a biomarker of exposure to EB/S in humans. It can also be used as a drug intermediate for the synthesis of antineoplastic compounds.
  • Analytical standard for research and analytical applications
  • Metabolite of ethylbenzene and styrene (EB/S)
  • Can be used as a biomarker of exposure to EB/S
  • Drug intermediate for the synthesis of antineoplastic compounds
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS