Learn More
Medchemexpress LLC Phenylglyoxylic acid sodium | 43165-51-1 | 99.9% | 172.12 | 500 MG
SDP
Catalog No. 5000366535
Encompass_Preferred
Phenylglyoxylic acid sodium is a metabolite of ethylbenzene and styrene (EB/S) and can be used as a biomarker of exposure to EB/S in humans. It can also be used as a drug intermediate for the synthesis of antineoplastic compounds. Studies show that Phenylglyoxylic acid sodium exhibits anti-inflammatory activity in rats in a formalin-induced paw swelling model.
  • Metabolite of ethylbenzene and styrene (EB/S)
  • Biomarker for exposure to EB/S in humans
  • Drug intermediate for the synthesis of antineoplastic compounds
  • Exhibits anti-inflammatory activity in rats
Catalog No. 50-003-66535 Supplier Medchemexpress LLC Supplier No. HYW010255A500MG
Phenylglyoxylic acid sodium is a metabolite of ethylbenzene and styrene (EB/S) and can be used as a biomarker of exposure to EB/S in humans. It can also be used as a drug intermediate for the synthesis of antineoplastic compounds. Studies show that Phenylglyoxylic acid sodium exhibits anti-inflammatory activity in rats in a formalin-induced paw swelling model.
  • Metabolite of ethylbenzene and styrene (EB/S)
  • Biomarker for exposure to EB/S in humans
  • Drug intermediate for the synthesis of antineoplastic compounds
  • Exhibits anti-inflammatory activity in rats